Product Details

SNP ID
rs115720142
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:138519080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAAGAAAACTGAGACCCAGAGAC[A/C]TTACACTGCTTTGGGATTGAACCCA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SPOPL PubMed Links

Gene Details

Gene
SPOPL
Gene Name
speckle type BTB/POZ protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001664.2 Intron NP_001001664.1
XM_005263655.4 Intron XP_005263712.1
XM_005263656.4 Intron XP_005263713.1
XM_005263657.4 Intron XP_005263714.1

View Full Product Details