Product Details

SNP ID
rs116151278
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:108327956 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAATAGAGAGGTACAATCTCTCA[A/G]TTAAACAAAGTGTTCAACCAGTTAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SULT1C2P1 PubMed Links

Gene Details

Gene
SULT1C2P1
Gene Name
sulfotransferase family 1C member 2 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details