Product Details

SNP ID
rs115907946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:68464135 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACAACAAAATGAACGAATAAAGA[C/T]ATCGCTTTTGCAACCTAAGAGTCAT
Phenotype
MIM: 611035
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APLF PubMed Links

Gene Details

Gene
APLF
Gene Name
aprataxin and PNKP like factor
There are no transcripts associated with this gene.

Gene
FBXO48
Gene Name
F-box protein 48
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024680.2 1011 UTR 3 NP_001019851.1
XM_005264407.2 1011 UTR 3 XP_005264464.1
XM_017004437.1 1011 UTR 3 XP_016859926.1

View Full Product Details