Product Details

SNP ID
rs116463148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:69659211 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTCAAGAAAGATTATAAACCATT[G/T]GGAAATAAATTAGAGATAGCATCTA
Phenotype
MIM: 106491
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ANXA4 PubMed Links

Gene Details

Gene
ANXA4
Gene Name
annexin A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001153.4 Intron NP_001144.1
NM_001320698.1 Intron NP_001307627.1
NM_001320700.1 Intron NP_001307629.1
NM_001320702.1 Intron NP_001307631.1
XM_017003943.1 Intron XP_016859432.1
XM_017003944.1 Intron XP_016859433.1

View Full Product Details