Product Details

SNP ID
rs114533892
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74455903 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATAATGAAGAGGAACCTATGGAA[A/G]GAGTCCCCCTTGAGCAGTACCGTGC
Phenotype
MIM: 616456 MIM: 601336 MIM: 606961
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INO80B PubMed Links

Gene Details

Gene
INO80B
Gene Name
INO80 complex subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031288.3 431 Missense Mutation AGA,GGA R113G NP_112578.2
Gene
INO80B-WBP1
Gene Name
INO80B-WBP1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
MOGS
Gene Name
mannosyl-oligosaccharide glucosidase
There are no transcripts associated with this gene.

Gene
WBP1
Gene Name
WW domain binding protein 1
There are no transcripts associated with this gene.

View Full Product Details