Product Details

SNP ID
rs114495106
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27381722 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGAGCTCTGCTGTGTTTCGGGG[C/T]TTGAAGCAAATGATGATGCAGGTCA
Phenotype
MIM: 605119 MIM: 605963 MIM: 613598
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPM1G PubMed Links

Gene Details

Gene
PPM1G
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_177983.2 1779 Silent Mutation AAA,AAG K506K NP_817092.1
Gene
SNX17
Gene Name
sorting nexin 17
There are no transcripts associated with this gene.

Gene
ZNF513
Gene Name
zinc finger protein 513
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201459.1 1779 Intron NP_001188388.1
NM_144631.5 1779 Intron NP_653232.3
XM_005264142.2 1779 Intron XP_005264199.1
XM_005264143.3 1779 Intron XP_005264200.1

View Full Product Details