Product Details

SNP ID
rs115600417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:69171466 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCCCTTCTCTCATCCTGCTGAA[C/T]TGTGCTCTGAGATTTCCCCTGTTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FRMD4B PubMed Links

Gene Details

Gene
FRMD4B
Gene Name
FERM domain containing 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015123.2 2974 UTR 3 NP_055938.1
XM_005264722.1 2974 UTR 3 XP_005264779.1
XM_005264723.1 2974 UTR 3 XP_005264780.1
XM_017005989.1 2974 UTR 3 XP_016861478.1
XM_017005990.1 2974 UTR 3 XP_016861479.1
XM_017005991.1 2974 UTR 3 XP_016861480.1
XM_017005992.1 2974 UTR 3 XP_016861481.1
XM_017005993.1 2974 UTR 3 XP_016861482.1
XM_017005994.1 2974 UTR 3 XP_016861483.1
XM_017005995.1 2974 UTR 3 XP_016861484.1
XM_017005996.1 2974 UTR 3 XP_016861485.1
XM_017005997.1 2974 UTR 3 XP_016861486.1

View Full Product Details