Product Details

SNP ID
rs114079758
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:69173653 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGACTGTATCCTAAAAAGAATCCT[A/C]AATAAACAAATGGATGGAAATAAGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FRMD4B PubMed Links

Gene Details

Gene
FRMD4B
Gene Name
FERM domain containing 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015123.2 Intron NP_055938.1
XM_005264722.1 Intron XP_005264779.1
XM_005264723.1 Intron XP_005264780.1
XM_017005989.1 Intron XP_016861478.1
XM_017005990.1 Intron XP_016861479.1
XM_017005991.1 Intron XP_016861480.1
XM_017005992.1 Intron XP_016861481.1
XM_017005993.1 Intron XP_016861482.1
XM_017005994.1 Intron XP_016861483.1
XM_017005995.1 Intron XP_016861484.1
XM_017005996.1 Intron XP_016861485.1
XM_017005997.1 Intron XP_016861486.1

View Full Product Details