Product Details

SNP ID
rs113328738
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:45224543 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACGCCCACCAGGCCACGTCACTGG[A/C]TAGATCAACTTGGAAAAGGTTCATA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TMEM158 PubMed Links

Gene Details

Gene
TMEM158
Gene Name
transmembrane protein 158 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015444.2 1780 UTR 3 NP_056259.2

View Full Product Details