Product Details

SNP ID
rs114869468
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:6861502 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCGGCGCGCGGCCAGGAGATGTA[C/T]GCCCCGCACTCAATCCGGATCGAGG
Phenotype
MIM: 604101
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRM7 PubMed Links

Gene Details

Gene
GRM7
Gene Name
glutamate metabotropic receptor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000844.3 388 Silent Mutation TAC,TAT Y38Y NP_000835.1
NM_181874.2 388 Silent Mutation TAC,TAT Y38Y NP_870989.1
XM_017006272.1 388 Intron XP_016861761.1
XM_017006273.1 388 Intron XP_016861762.1

View Full Product Details