Product Details

SNP ID
rs116840785
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:8733961 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGATTGACCTGGTGAACCGAGAC[A/C]CCAAGAACATTAACGAGGACATAGT
Phenotype
MIM: 601253
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CAV3 PubMed Links

Gene Details

Gene
CAV3
Gene Name
caveolin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001234.4 162 Missense Mutation ACC,CCC T29P NP_001225.1
NM_033337.2 162 Missense Mutation ACC,CCC T29P NP_203123.1
Gene
SSUH2
Gene Name
ssu-2 homolog (C. elegans)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256748.1 162 Intron NP_001243677.1
NM_001256749.1 162 Intron NP_001243678.1
NM_015931.2 162 Intron NP_057015.1
XM_011533774.1 162 Intron XP_011532076.1
XM_017006510.1 162 Intron XP_016861999.1
XM_017006511.1 162 Intron XP_016862000.1
XM_017006512.1 162 Intron XP_016862001.1
XM_017006513.1 162 Intron XP_016862002.1
XM_017006514.1 162 Intron XP_016862003.1
XM_017006515.1 162 Intron XP_016862004.1
XM_017006516.1 162 Intron XP_016862005.1
XM_017006517.1 162 Intron XP_016862006.1
XM_017006518.1 162 Intron XP_016862007.1
XM_017006519.1 162 Intron XP_016862008.1
XM_017006520.1 162 Intron XP_016862009.1
XM_017006521.1 162 Intron XP_016862010.1
XM_017006522.1 162 Intron XP_016862011.1
XM_017006523.1 162 Intron XP_016862012.1
XM_017006524.1 162 Intron XP_016862013.1
XM_017006525.1 162 Intron XP_016862014.1
XM_017006526.1 162 Intron XP_016862015.1
XM_017006527.1 162 Intron XP_016862016.1
XM_017006528.1 162 Intron XP_016862017.1
XM_017006529.1 162 Intron XP_016862018.1
XM_017006530.1 162 Intron XP_016862019.1
XM_017006531.1 162 Intron XP_016862020.1
XM_017006532.1 162 Intron XP_016862021.1
XM_017006533.1 162 Intron XP_016862022.1
XM_017006534.1 162 Intron XP_016862023.1
XM_017006535.1 162 Intron XP_016862024.1
XM_017006536.1 162 Intron XP_016862025.1
XM_017006537.1 162 Intron XP_016862026.1
XM_017006538.1 162 Intron XP_016862027.1
XM_017006539.1 162 Intron XP_016862028.1
XM_017006540.1 162 Intron XP_016862029.1
XM_017006541.1 162 Intron XP_016862030.1
XM_017006542.1 162 Intron XP_016862031.1

View Full Product Details