Product Details

SNP ID
rs112200203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177458157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCAAGGGCTGAGGGCAGAGTTGG[C/G]GTCTGGGGGGCAGCCAGGGACTCCC
Phenotype
MIM: 126660
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DBN1 PubMed Links

Gene Details

Gene
DBN1
Gene Name
drebrin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004395.3 1965 Silent Mutation ACC,ACG T559T NP_004386.2
NM_080881.2 1965 Silent Mutation ACC,ACG T561T NP_543157.1
XM_005265827.3 1965 Silent Mutation ACC,ACG T605T XP_005265884.1
XM_006714826.2 1965 Silent Mutation ACC,ACG T542T XP_006714889.1
XM_011534446.2 1965 Silent Mutation ACC,ACG T607T XP_011532748.1
XM_011534447.2 1965 Silent Mutation ACC,ACG T594T XP_011532749.1
XM_017009139.1 1965 Silent Mutation ACC,ACG T607T XP_016864628.1
XM_017009140.1 1965 Silent Mutation ACC,ACG T561T XP_016864629.1
Gene
PRR7
Gene Name
proline rich 7 (synaptic)
There are no transcripts associated with this gene.

View Full Product Details