Product Details

SNP ID
rs116305151
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:179679095 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCAAAACTGCTGCAGCGTCTGCC[A/C]CAGGCGGCTGGCATGGCTCGTAGCC
Phenotype
MIM: 114217
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CANX PubMed Links

Gene Details

Gene
CANX
Gene Name
calnexin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024649.1 1126 Intron NP_001019820.1
NM_001746.3 1126 Intron NP_001737.1
XM_011534664.1 1126 Intron XP_011532966.1
XM_011534665.2 1126 Intron XP_011532967.1
Gene
CBY3
Gene Name
chibby family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164444.1 1126 Missense Mutation GGG,TGG G73W NP_001157916.1
XM_017009726.1 1126 Missense Mutation GGG,TGG G72W XP_016865215.1
Gene
LOC105377763
Gene Name
uncharacterized LOC105377763
There are no transcripts associated with this gene.

View Full Product Details