Product Details

SNP ID
rs116056653
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:36434485 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGTTTGCTTTTTAAAACTTAAC[A/G]CCTGAGACTTGGTTCTTATGAATTG
Phenotype
MIM: 604036
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCNA1 PubMed Links

Gene Details

Gene
CCNA1
Gene Name
cyclin A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111045.1 Intron NP_001104515.1
NM_001111046.1 Intron NP_001104516.1
NM_001111047.1 Intron NP_001104517.1
NM_003914.3 Intron NP_003905.1
XM_011535294.2 Intron XP_011533596.1
XM_011535295.2 Intron XP_011533597.1
XM_011535296.2 Intron XP_011533598.1

View Full Product Details