Product Details

SNP ID
rs115325365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:44943481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGATGGTGTGTTCTTGGTTCGAA[A/T]AACGTTTGATAGTTGTGATAATCTT
Phenotype
MIM: 604354
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NUFIP1 PubMed Links

Gene Details

Gene
NUFIP1
Gene Name
NUFIP1, FMR1 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012345.2 Intron NP_036477.2

View Full Product Details