Product Details

SNP ID
rs117241647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:45545396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACTCTGTTGCCATGCCTTGTTG[A/G]TTTCCTTCATATAATTTACGAATAG
Phenotype
MIM: 606975
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COG3 PubMed Links

Gene Details

Gene
COG3
Gene Name
component of oligomeric golgi complex 3
There are no transcripts associated with this gene.

Gene
ERICH6B
Gene Name
glutamate rich 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182542.2 Intron NP_872348.2
XM_011534965.2 Intron XP_011533267.1
XM_017020418.1 Intron XP_016875907.1

View Full Product Details