Product Details

SNP ID
rs114638218
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:46341025 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGATCTAATCTGAAGTTGTGGCC[A/G]ATAATTCTCACCCAGGTCAACACAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA0226L PubMed Links

Gene Details

Gene
KIAA0226L
Gene Name
KIAA0226 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286761.1 Intron NP_001273690.1
NM_001286762.1 Intron NP_001273691.1
NM_001286763.1 Intron NP_001273692.1
NM_001286764.1 Intron NP_001273693.1
NM_001286765.1 Intron NP_001273694.1
NM_001286766.1 Intron NP_001273695.1
NM_025113.3 Intron NP_079389.2

View Full Product Details