Product Details

SNP ID
rs113980525
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:78601914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCGAGTCGCAGATGGACGCCAGG[C/T]CCGCTGCGCCCACCACGGCCGCCGC
Phenotype
MIM: 601632
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC107987188 PubMed Links

Gene Details

Gene
LOC107987188
Gene Name
uncharacterized LOC107987188
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017020899.1 995 Intron XP_016876388.1
Gene
POU4F1
Gene Name
POU class 4 homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006237.3 995 Missense Mutation GAC,GGC D254G NP_006228.3
Gene
RNF219-AS1
Gene Name
RNF219 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details