Product Details
- SNP ID
-
rs115398523
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:80116447 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAATCTTTTGTGGGACAGCCAGAC[C/T]GTTTTCAGGCACACATGAAGACTGG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ADNP2
PubMed Links
Gene Details
- Gene
- ADNP2
- Gene Name
- ADNP homeobox 2
View Full Product Details