Product Details

SNP ID
rs118066714
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:59903275 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCATTCTGAGAACACTGAAATGTT[A/G]TACTCAGTGTTGATTTCTTCGGTCA
Phenotype
MIM: 604959
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PMAIP1 PubMed Links

Gene Details

Gene
PMAIP1
Gene Name
phorbol-12-myristate-13-acetate-induced protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021127.2 905 UTR 3 NP_066950.1

View Full Product Details