Product Details

SNP ID
rs116632144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:132609287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAGTTAGATCCATGGTGCTTTAT[C/T]CCTTGAAATTTCTTTCCAGGAGGAT
Phenotype
MIM: 604849
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAAR2 PubMed Links

Gene Details

Gene
TAAR2
Gene Name
trace amine associated receptor 2
There are no transcripts associated with this gene.

Gene
TAAR3
Gene Name
trace amine associated receptor 3 (gene/pseudogene)
There are no transcripts associated with this gene.

View Full Product Details