Product Details

SNP ID
rs117582035
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:169739127 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAAATCATACTGATTATCACAGG[C/T]ATTGCTTGCCTCCCACAAGTGCACA
Phenotype
MIM: 186977
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TCTE3 PubMed Links

Gene Details

Gene
TCTE3
Gene Name
t-complex-associated-testis-expressed 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006715554.3 Intron XP_006715617.1
XM_011536093.2 Intron XP_011534395.1

View Full Product Details