Product Details

SNP ID
rs111529266
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:58641672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTTCAGTGAGTGTTTATCCAGT[C/T]GTCATTCCAGCACCTGCTTTTGCAG
Phenotype
MIM: 607861
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DACT1 PubMed Links

Gene Details

Gene
DACT1
Gene Name
dishevelled binding antagonist of beta catenin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079520.1 723 Missense Mutation CGT,TGT R187C NP_001072988.1
NM_016651.5 723 Missense Mutation CGT,TGT R187C NP_057735.2
XM_006720167.3 723 UTR 5 XP_006720230.1

View Full Product Details