Product Details

SNP ID
rs115921791
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:69777364 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAAGGGAGAGAGAATCGGCATTT[A/G]CTTGCTAGTATAATGTAACAAATTC
Phenotype
MIM: 182396 MIM: 600914
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100289511 PubMed Links

Gene Details

Gene
LOC100289511
Gene Name
uncharacterized LOC100289511
There are no transcripts associated with this gene.

Gene
SLC10A1
Gene Name
solute carrier family 10 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003049.3 Intron NP_003040.1
Gene
SRSF5
Gene Name
serine and arginine rich splicing factor 5
There are no transcripts associated with this gene.

View Full Product Details