Product Details

SNP ID
rs116115069
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:70729171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACCTCAAGGTTATTGTACATCAG[C/T]GGCTCCCTGGGCACCCTATAAATAG
Phenotype
MIM: 600136
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MAP3K9 PubMed Links

Gene Details

Gene
MAP3K9
Gene Name
mitogen-activated protein kinase kinase kinase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284230.1 4550 UTR 3 NP_001271159.1
NM_001284231.1 4550 UTR 3 NP_001271160.1
NM_001284232.1 4550 UTR 3 NP_001271161.1
NM_033141.3 4550 UTR 3 NP_149132.2
XM_005267683.4 4550 UTR 3 XP_005267740.1
XM_011536788.2 4550 UTR 3 XP_011535090.1
XM_011536792.1 4550 UTR 3 XP_011535094.1
XM_011536794.2 4550 UTR 3 XP_011535096.1

View Full Product Details