Product Details

SNP ID
rs116434191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77277454 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGGCCAGGGGACCAACCATCCC[A/G]TAAGCCAGAGGGTGGAAGAGGTAGA
Phenotype
MIM: 605304 MIM: 607439
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NGB PubMed Links

Gene Details

Gene
NGB
Gene Name
neuroglobin
There are no transcripts associated with this gene.

Gene
POMT2
Gene Name
protein O-mannosyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013382.5 2478 Silent Mutation NP_037514.2
XM_011536675.2 2478 Silent Mutation XP_011534977.1
XM_011536676.2 2478 Silent Mutation XP_011534978.1
XM_011536677.2 2478 Silent Mutation XP_011534979.1
XM_011536679.2 2478 Silent Mutation XP_011534981.1
XM_017021244.1 2478 Silent Mutation XP_016876733.1
XM_017021245.1 2478 Silent Mutation XP_016876734.1

View Full Product Details