Product Details

SNP ID
rs117474357
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:93999320 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGACTCTGGCCAGAAAGGCTCAG[A/C]CTTCATTCTCCCCCAGATACCTTCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC00521 PubMed Links

Gene Details

Gene
LINC00521
Gene Name
long intergenic non-protein coding RNA 521
There are no transcripts associated with this gene.

View Full Product Details