Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001199427.1 | Intron | NP_001186356.1 | |||
NM_003300.3 | Intron | NP_003291.2 | |||
NM_145725.2 | Intron | NP_663777.1 | |||
NM_145726.2 | Intron | NP_663778.1 | |||
XM_011537116.2 | Intron | XP_011535418.1 | |||
XM_011537117.2 | Intron | XP_011535419.1 | |||
XM_011537118.2 | Intron | XP_011535420.1 | |||
XM_017021617.1 | Intron | XP_016877106.1 | |||
XM_017021618.1 | Intron | XP_016877107.1 | |||
XM_017021619.1 | Intron | XP_016877108.1 | |||
XM_017021620.1 | Intron | XP_016877109.1 |