Product Details

SNP ID
rs112197784
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:104989669 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGAGCATTGAAAGACAGACAGC[A/G]CTCGAGTGGATTAGGCCGCAGTGGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C14orf79 PubMed Links

Gene Details

Gene
C14orf79
Gene Name
chromosome 14 open reading frame 79
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174891.3 Intron NP_777551.2
XM_005267317.3 Intron XP_005267374.1
XM_005267318.3 Intron XP_005267375.1

View Full Product Details