Product Details

SNP ID
rs115807156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105209950 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCAGGGGTGGGGGTGTCTGCCT[C/T]GGACGAGGCAGGTATCTGGATGCAG
Phenotype
MIM: 604902
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BRF1 PubMed Links

Gene Details

Gene
BRF1
Gene Name
BRF1, RNA polymerase III transcription initiation factor 90 kDa subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242786.1 3288 UTR 3 NP_001229715.1
NM_001242787.1 3288 UTR 3 NP_001229716.1
NM_001242788.1 3288 UTR 3 NP_001229717.1
NM_001242789.1 3288 UTR 3 NP_001229718.1
NM_001242790.1 3288 Intron NP_001229719.1
NM_001519.3 3288 UTR 3 NP_001510.2
NM_145685.2 3288 UTR 3 NP_663718.1
XM_005267561.3 3288 UTR 3 XP_005267618.1
XM_005267563.3 3288 UTR 3 XP_005267620.1
XM_006720123.2 3288 UTR 3 XP_006720186.1
XM_011536672.2 3288 Intron XP_011534974.1
XM_011536673.1 3288 UTR 3 XP_011534975.1
XM_011536674.1 3288 UTR 3 XP_011534976.1

View Full Product Details