Product Details

SNP ID
rs115541869
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105602654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCAACCAGCTTGCTTCTGACTCA[A/G]CTCCTCTTAGCCAGGTGAGAATGGT
Phenotype
MIM: 146910 MIM: 147070 MIM: 147010
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IGH PubMed Links

Gene Details

Gene
IGH
Gene Name
immunoglobulin heavy locus
There are no transcripts associated with this gene.

View Full Product Details