Product Details
- SNP ID
-
rs112222496
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52898867 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAATGGCCAGCTCTCCACGCTGCTC[A/G]GCATCTGCAATGGCGGCCTCCAGGG
- Phenotype
-
MIM: 148060
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT8
PubMed Links
Gene Details
- Gene
- KRT8
- Gene Name
- keratin 8
View Full Product Details