Product Details

SNP ID
rs115871774
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55761819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATCTGAAATCAGGTAGGCTCCTC[A/G]GGTGTCTTTCCCCCAGGAAGGACTT
Phenotype
MIM: 603936 MIM: 610049
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GDF11 PubMed Links

Gene Details

Gene
GDF11
Gene Name
growth differentiation factor 11
There are no transcripts associated with this gene.

Gene
SARNP
Gene Name
SAP domain containing ribonucleoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033082.3 Intron NP_149073.1

View Full Product Details