Product Details

SNP ID
rs117611147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:64718856 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACTGTTGGAAATTGCAAAGTCAG[C/G]ACATGTAAACTGGAAGTCTTGTGCT
Phenotype
MIM: 607664
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GNS PubMed Links

Gene Details

Gene
GNS
Gene Name
glucosamine (N-acetyl)-6-sulfatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002076.3 Intron NP_002067.1

View Full Product Details