Product Details

SNP ID
rs112721024
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:72094087 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTGGGTTTGCAGTGGAGTCTGT[G/T]GTTGGTGAGCCTCTTATCAGGACTG
Phenotype
MIM: 606950
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TRHDE PubMed Links

Gene Details

Gene
TRHDE
Gene Name
thyrotropin releasing hormone degrading enzyme
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013381.2 Intron NP_037513.1
XM_005268819.4 Intron XP_005268876.1
XM_011538248.2 Intron XP_011536550.1
XM_017019243.1 Intron XP_016874732.1
XM_017019244.1 Intron XP_016874733.1

View Full Product Details