Product Details

SNP ID
rs117659653
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:106991506 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAAAGTTTAAAACATATACTGTC[C/T]GTGTTAACAGAGGCTACAATACCAA
Phenotype
MIM: 601933 MIM: 616929
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRY1 PubMed Links

Gene Details

Gene
CRY1
Gene Name
cryptochrome circadian clock 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004075.4 3153 UTR 3 NP_004066.1
XM_017018832.1 3153 UTR 3 XP_016874321.1
Gene
MTERF2
Gene Name
mitochondrial transcription termination factor 2
There are no transcripts associated with this gene.

View Full Product Details