Product Details
- SNP ID
-
rs111364787
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:116778953 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGTTGCGGAGCCAGGCCTGCCTCA[G/T]AGCCCTCTGTCTTCATGTCATTGAG
- Phenotype
-
MIM: 603901
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
BUD13
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75198898] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BUD13
- Gene Name
- BUD13 homolog
There are no transcripts associated with this gene.
- Gene
- ZPR1
- Gene Name
- ZPR1 zinc finger
View Full Product Details