Product Details

SNP ID
rs113034222
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125677870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGCTCACCTGAAGGCTGTTCACC[C/T]GAGGCCTGCTCACCAGAAGGCTGTT
Phenotype
MIM: 102525 MIM: 603078
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACRV1 PubMed Links

Gene Details

Gene
ACRV1
Gene Name
acrosomal vesicle protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001612.5 598 Silent Mutation TCA,TCG S160S NP_001603.1
NM_020069.4 598 Silent Mutation TCA,TCG S160S NP_064454.1
NM_020107.4 598 Silent Mutation TCA,TCG S105S NP_064492.1
NM_020108.4 598 Silent Mutation TCA,TCG S90S NP_064493.1
Gene
CHEK1
Gene Name
checkpoint kinase 1
There are no transcripts associated with this gene.

View Full Product Details