Product Details

SNP ID
rs115805775
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:128461393 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAACAGTGCTCCTACGTTTACTGT[C/T]GTCCAAAACCAGGGATGCAAAATAC
Phenotype
MIM: 164720
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ETS1 PubMed Links

Gene Details

Gene
ETS1
Gene Name
ETS proto-oncogene 1, transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143820.1 2285 UTR 3 NP_001137292.1
NM_001162422.1 2285 UTR 3 NP_001155894.1
NM_005238.3 2285 UTR 3 NP_005229.1
XM_011542649.2 2285 UTR 3 XP_011540951.1
XM_011542650.1 2285 UTR 3 XP_011540952.1
XM_011542651.1 2285 UTR 3 XP_011540953.1
XM_011542652.1 2285 UTR 3 XP_011540954.1
XM_017017314.1 2285 UTR 3 XP_016872803.1
XM_017017315.1 2285 UTR 3 XP_016872804.1
XM_017017316.1 2285 UTR 3 XP_016872805.1
XM_017017317.1 2285 UTR 3 XP_016872806.1

View Full Product Details