Product Details

SNP ID
rs117834634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:50444756 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAATGTTAGTCCTGATAAAAATT[A/T]AAAACCATAAAAGAGGTAACAAATT
Phenotype
MIM: 615383
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FIGNL1 PubMed Links

Gene Details

Gene
FIGNL1
Gene Name
fidgetin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042762.2 2911 UTR 3 NP_001036227.1
NM_001287492.1 2911 UTR 3 NP_001274421.1
NM_001287493.1 2911 UTR 3 NP_001274422.1
NM_001287494.1 2911 UTR 3 NP_001274423.1
NM_001287495.1 2911 UTR 3 NP_001274424.1
NM_001287496.1 2911 UTR 3 NP_001274425.1
NM_022116.4 2911 UTR 3 NP_071399.2
XM_011515470.2 2911 UTR 3 XP_011513772.1
XM_017012501.1 2911 UTR 3 XP_016867990.1
XM_017012502.1 2911 UTR 3 XP_016867991.1

View Full Product Details