Product Details

SNP ID
rs116551965
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:102237742 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATAAATTGAAGAACATTCCCTTC[C/T]AGGACAGAAAATAATTCTAAAAATT
Phenotype
MIM: 609013
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
SLCO4C1 PubMed Links

Gene Details

Gene
SLCO4C1
Gene Name
solute carrier organic anion transporter family member 4C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_180991.4 Intron NP_851322.3
XM_011543370.1 Intron XP_011541672.1
XM_011543372.1 Intron XP_011541674.1

View Full Product Details