Product Details

SNP ID
rs113641537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48460730 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGAGGATGAGCGGGTCCTTCTG[C/G]CTGTACATGTGCAGCATCTCCGCCT
Phenotype
MIM: 300649
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC38A5 PubMed Links

Gene Details

Gene
SLC38A5
Gene Name
solute carrier family 38 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033518.3 1165 Missense Mutation AGC,AGG S329R NP_277053.2
XM_005272694.3 1165 Missense Mutation AGC,AGG S376R XP_005272751.2
XM_005272695.4 1165 Missense Mutation AGC,AGG S376R XP_005272752.3
XM_005272697.2 1165 Missense Mutation AGC,AGG S335R XP_005272754.2
XM_005272698.4 1165 Missense Mutation AGC,AGG S329R XP_005272755.2
XM_006724569.3 1165 Missense Mutation AGC,AGG S329R XP_006724632.1
XM_017029960.1 1165 Missense Mutation AGC,AGG S335R XP_016885449.1
XM_017029961.1 1165 Missense Mutation AGC,AGG S329R XP_016885450.1

View Full Product Details