Product Details

SNP ID
rs111610360
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102854367 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTTCTCACGGAGAAGGTGACCAC[C/T]TGTGGTACTGACGTAATCGCGCTCA
Phenotype
MIM: 616600
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BORCS7 PubMed Links

Gene Details

Gene
BORCS7
Gene Name
BLOC-1 related complex subunit 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136200.1 158 Silent Mutation ACC,ACT T27T NP_001129672.1
NM_144591.3 158 Silent Mutation ACC,ACT T27T NP_653192.2
Gene
BORCS7-ASMT
Gene Name
BORCS7-ASMT readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details