Product Details

SNP ID
rs112032940
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:75103972 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGGAGTAGCAGGCTCTAGGGCC[A/G]ACCCCACGCTGGGCTTCCACCATCT
Phenotype
MIM: 613191 MIM: 611575
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUSP13 PubMed Links

Gene Details

Gene
DUSP13
Gene Name
dual specificity phosphatase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007271.1 330 UTR 3 NP_001007272.1
NM_001007272.1 330 Intron NP_001007273.1
NM_001007273.1 330 Silent Mutation GTC,GTT V35V NP_001007274.1
NM_001320842.1 330 Silent Mutation GTC,GTT V35V NP_001307771.1
NM_001320843.1 330 Intron NP_001307772.1
NM_016364.3 330 Intron NP_057448.3
XM_005269883.3 330 Silent Mutation GTC,GTT V161V XP_005269940.1
XM_005269884.4 330 Intron XP_005269941.3
XM_005269887.1 330 Silent Mutation GTC,GTT V35V XP_005269944.1
XM_005269890.1 330 Intron XP_005269947.1
XM_011539853.1 330 Silent Mutation GTC,GTT V71V XP_011538155.1
XM_011539854.2 330 Intron XP_011538156.1
XM_011539855.1 330 Intron XP_011538157.1
XM_011539856.2 330 Intron XP_011538158.1
XM_017016313.1 330 Intron XP_016871802.1
XM_017016314.1 330 Silent Mutation GTC,GTT V161V XP_016871803.1
Gene
SAMD8
Gene Name
sterile alpha motif domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174156.1 330 Intron NP_001167627.1
NM_144660.2 330 Intron NP_653261.1
XM_005269541.4 330 Intron XP_005269598.1
XM_011539311.1 330 Intron XP_011537613.1
XM_011539312.2 330 Intron XP_011537614.1
XM_017015738.1 330 Intron XP_016871227.1
XM_017015739.1 330 Intron XP_016871228.1

View Full Product Details