Product Details

SNP ID
rs113581631
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:5393916 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGGCCATGTACTTCCCAAGCCG[A/G]GGATCACACTTGACCAGCTGGTTGG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TUBAL3 PubMed Links

Gene Details

Gene
TUBAL3
Gene Name
tubulin alpha like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171864.1 980 Silent Mutation CCC,CCT P274P NP_001165335.1
NM_024803.2 980 Silent Mutation CCC,CCT P314P NP_079079.1

View Full Product Details