Product Details

SNP ID
rs116325368
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:106574740 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACTTTGAGAACAACTGGCCAAGG[C/T]CCTAAGAACAATGAGGTCCTCCTCT
Phenotype
MIM: 606283
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SORCS1 PubMed Links

Gene Details

Gene
SORCS1
Gene Name
sortilin related VPS10 domain containing receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013031.2 6530 UTR 3 NP_001013049.1
NM_001206569.1 6530 Intron NP_001193498.1
NM_001206570.1 6530 UTR 3 NP_001193499.1
NM_001206571.1 6530 Intron NP_001193500.1
NM_001206572.1 6530 UTR 3 NP_001193501.1
NM_052918.4 6530 UTR 3 NP_443150.3
XM_011539199.2 6530 Intron XP_011537501.1
XM_011539201.2 6530 Intron XP_011537503.1
XM_017015614.1 6530 Intron XP_016871103.1
XM_017015615.1 6530 Intron XP_016871104.1
XM_017015616.1 6530 Intron XP_016871105.1
XM_017015617.1 6530 Intron XP_016871106.1
XM_017015618.1 6530 Intron XP_016871107.1

View Full Product Details