Product Details

SNP ID
rs137965903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:119651700 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGAGCGCCGCCACCCACTCGCCC[A/G]TGATGCAGGTGGCGTCCGGCAACGG
Phenotype
MIM: 603883
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BAG3 PubMed Links

Gene Details

Gene
BAG3
Gene Name
BCL2 associated athanogene 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004281.3 331 Missense Mutation ATG,GTG M9V NP_004272.2
XM_005270287.1 331 Missense Mutation ATG,GTG M9V XP_005270344.1

View Full Product Details