Product Details

SNP ID
rs138355284
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:110498229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGGCCCTATCTGGCCTTCGCTGC[C/T]TCGAACGTGCGCGGCTCGCTCAACG
Phenotype
MIM: 603069
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP5 PubMed Links

Gene Details

Gene
DUSP5
Gene Name
dual specificity phosphatase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004419.3 363 Silent Mutation GCC,GCT A36A NP_004410.3

View Full Product Details