Product Details

SNP ID
rs138631375
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133326944 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGGCCTTGTACATGATCAGCAC[A/G]CAGCCCAGTAGCGCCATGGCGAAGG
Phenotype
MIM: 604647
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CALY PubMed Links

Gene Details

Gene
CALY
Gene Name
calcyon neuron specific vesicular protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321617.1 1596 UTR 5 NP_001308546.1
NM_015722.3 1596 Silent Mutation TGC,TGT C98C NP_056537.1
XM_005252685.4 1596 Silent Mutation TGC,TGT C131C XP_005252742.2
XM_011539836.2 1596 Silent Mutation TGC,TGT C131C XP_011538138.1
XM_011539838.2 1596 Silent Mutation TGC,TGT C11C XP_011538140.1

View Full Product Details