Product Details

SNP ID
rs138990977
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:17321086 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATTTTTACACAGTTCCACTGCA[A/G]CACTTGTGATCATCAAGCCGGTGGA
Phenotype
MIM: 610139
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ST8SIA6 PubMed Links

Gene Details

Gene
ST8SIA6
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004470.1 981 Missense Mutation GCT,GTT A330V NP_001004470.1
XM_017016174.1 981 Missense Mutation GCT,GTT A300V XP_016871663.1
XM_017016175.1 981 Intron XP_016871664.1

View Full Product Details